Search
Search

TNFSF10| Gene Symbol: TNFSF10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001190942.1 | NP_001177871.1 | 1 | 176 | ||
| NM_001190943.1 | NP_001177872.1 | 1 | 144 | |||
| NM_003810.3 | NP_003801.1 | 1 | 176 | |||
| NR_033994.1 | 1 | 176 | ||||
| GenBank mRNA | AK296085.1 | - | 1 | 144 | ||
| AK309200.1 | - | 1 | 136 | |||
| AK312742.1 | - | 1 | 144 | |||
| BC009795.1 | - | 1 | 73 | |||
| BC032722.1 | - | 1 | 128 | 5504670 | ||
| BT019564.1 | - | 1 | 53 | |||
| CR456895.1 | - | 1 | 53 | |||
| DA041872.1 | - | 1 | 136 | |||
| DA969653.1 | - | 1 | 176 | |||
| DQ848564.1 | - | 1 | 53 | |||
| EU183231.1 | - | 1 | 53 | |||
| U37518.1 | - | 1 | 140 | |||
| U57059.1 | - | 1 | 143 | |||