Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other RNF8 SIRNA Assays ›
          Gene Symbol:
          RNF8
          Gene Name:
          ring finger protein 8
          Gene Aliases:
          hRNF8
          Chromosome Location:
          Chr.6: 37353972 - 37394738 on Build GRCh38
          UniGene ID:
          Hs.485278
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID HSS113323
          Size 20 nmol
          Availability Made To Order
          Catalog # 1299001
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: RNF8
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_003958.3 NP_003949.1 3, 4 1178
          NM_183078.2 NP_898901.1 3, 4 1178
          NR_046399.1 3, 4 1284
          XM_006715241.2 XP_006715304.1 3, 4 1178
          XM_006715242.2 XP_006715305.1 3, 4 1178
          XM_017011462.1 XP_016866951.1 3, 4 1029
          XM_017011463.1 XP_016866952.1 3, 4 1034
          XM_017011464.1 XP_016866953.1 3, 4 1183
          GenBank mRNA AB014546.1 - 3, 4 1097
          AF334675.1 - 3, 4 985
          AK022075.1 - 3, 4 1079
          AK222765.1 - 3, 4 1096
          AK298319.1 - 3, 4 1202
          BC007517.2 - 3, 4 1107 3028746
          BT007446.1 - 3, 4 985

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          ubiquitin-protein ligase protein modifying enzyme

          Gene Ontology Categories:

          Function(s) Process(es)

          double-strand break repair
          double-strand break repair via nonhomologous end joining
          ubiquitin-dependent protein catabolic process
          cellular response to DNA damage stimulus
          mitotic nuclear division
          spermatid development
          response to ionizing radiation
          histone H2A ubiquitination
          histone H2B ubiquitination
          negative regulation of transcription elongation from RNA polymerase II promoter
          interstrand cross-link repair
          histone exchange
          isotype switching
          positive regulation of DNA repair
          cell division
          protein autoubiquitination
          protein K63-linked ubiquitination
          histone H2A K63-linked ubiquitination
          protein K48-linked ubiquitination
          chromatin binding
          ubiquitin-protein transferase activity
          protein binding
          zinc ion binding
          ligase activity
          ubiquitin protein ligase binding
          histone binding
          protein homodimerization activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-ctdjb:80/100.66.76.55:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0