Search
Search

CCNE2| Gene Symbol: CCNE2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_057749.2 | NP_477097.1 | 9 | 832 | ||
| XM_011517366.2 | XP_011515668.1 | 8 | 1157 | |||
| XM_017013957.1 | XP_016869446.1 | 8 | 1157 | |||
| XM_017013958.1 | XP_016869447.1 | 9 | 974 | |||
| XM_017013959.1 | XP_016869448.1 | 9 | 1076 | |||
| GenBank mRNA | AF091433.1 | - | 8 | 765 | ||
| AF102778.1 | - | 7 | 728 | |||
| AF106690.1 | - | 7 | 728 | |||
| AF112857.1 | - | 7 | 593 | |||
| BC007015.1 | - | 7 | 567 | 3997513 | ||
| BC020729.1 | - | 9 | 779 | 4774554 | ||