Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other TSPOAP1 SIRNA Assays ›
Gene Symbol:
TSPOAP1
Gene Name:
TSPO associated protein 1
Gene Aliases:
BZRAP1, PBR-IP, PRAX-1, PRAX1, RIM-BP1, RIMBP1
Chromosome Location:
Chr.17: 58301227 - 58329030 on Build GRCh38
UniGene ID:
Hs.112499
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
HSS113727
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Contact Us
Your Price
Online offer:
3062.0
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: TSPOAP1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001261835.1
NP_001248764.1
16
2885
NM_004758.3
NP_004749.2
16
2885
NM_024418.2
NP_077729.1
15
2705
XM_005257807.2
XP_005257864.1
16
2889
XM_006722173.2
XP_006722236.1
16
2889
XM_006722174.2
XP_006722237.1
15
2868
XM_006722175.2
XP_006722238.1
15
2709
XM_006722176.2
XP_006722239.1
16
2889
XM_006722177.3
XP_006722240.1
12
1157
XM_006722178.3
XP_006722241.1
11
1078
XM_011525471.2
XP_011523773.1
4
676
XM_011525472.1
XP_011523774.1
6
1378
XM_017025326.1
XP_016880815.1
16
2889
XM_017025327.1
XP_016880816.1
17
2037
XM_017025328.1
XP_016880817.1
14
2580
XM_017025329.1
XP_016880818.1
16
2889
XM_017025330.1
XP_016880819.1
16
2889
XM_017025331.1
XP_016880820.1
16
2889
XM_017025332.1
XP_016880821.1
15
2709
XM_017025333.1
XP_016880822.1
16
2889
XM_017025334.1
XP_016880823.1
15
2709
GenBank mRNA
AB014512.2
-
15
2674
AF039571.1
-
16
2211
BC146852.1
-
16
2178
9056780
BC150279.1
-
15
2674
9007261
BC171787.1
-
16
2178
9054441
BF934025.1
-
3
202
KJ534782.1
-
16
2014
KJ534783.1
-
16
2014
KJ534785.1
-
16
2014
KJ534787.1
-
15
1834
KJ534788.1
-
16
2014
KJ534789.1
-
13
1597
KJ534790.1
-
14
1726
KJ535016.1
-
16
2014
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
neurotransmitter secretion
biological_process
glutamate secretion
protein binding
benzodiazepine receptor binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-8jdkq:80/100.66.75.107:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0