Search
Search

SOX13| Gene Symbol: SOX13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_005686.2 | NP_005677.2 | 2 | 655 | ||
| XM_005245623.2 | XP_005245680.1 | 2 | 677 | |||
| GenBank mRNA | AF083105.1 | - | 2 | 117 | ||
| AF098915.1 | - | 1 | 60 | |||
| AF116571.1 | - | 2 | 168 | |||
| AK304192.1 | - | 2 | 118 | |||
| AK314197.1 | - | 2 | 151 | |||
| BC106038.1 | - | 2 | 674 | 6502746 | ||
| BQ935858.1 | - | 2 | 646 | |||