Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other GDA SIRNA Assays ›
          Gene Symbol:
          GDA
          Gene Name:
          guanine deaminase
          Gene Aliases:
          CYPIN, GUANASE, NEDASIN
          Chromosome Location:
          Chr.9: 72114595 - 72256045 on Build GRCh38
          UniGene ID:
          Hs.494163
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID HSS114360
          Size 20 nmol
          Availability Made To Order
          Catalog # 1299001
          Price Contact Us
          Your Price
          Online offer:
          3062.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: GDA
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001242505.2 NP_001229434.1 4, 5 675
          NM_001242506.2 NP_001229435.1 5, 6 716
          NM_001242507.2 NP_001229436.1 4, 5 582
          NM_004293.4 NP_004284.1 4, 5 675
          XM_005252317.1 XP_005252374.1 4, 5 673
          XM_011519213.1 XP_011517515.1 5, 6 787
          XM_011519214.1 XP_011517516.1 5, 6 787
          XM_011519215.1 XP_011517517.1 5, 6 787
          XM_011519216.1 XP_011517518.1 5, 6 787
          XM_011519217.1 XP_011517519.1 5, 6 787
          XM_011519218.2 XP_011517520.1 5, 6 672
          XM_011519219.1 XP_011517521.1 5, 6 785
          XM_011519220.1 XP_011517522.1 4, 5 671
          XM_011519221.1 XP_011517523.1 4, 5 582
          XM_011519222.1 XP_011517524.1 5, 6 787
          XM_011519223.2 XP_011517525.1 2, 3 152
          XM_017015333.1 XP_016870822.1 5, 6 787
          XM_017015334.1 XP_016870823.1 5, 6 787
          XM_017015335.1 XP_016870824.1 5, 6 787
          XM_017015336.1 XP_016870825.1 4, 5 673
          XM_017015337.1 XP_016870826.1 4, 5 673
          XM_017015338.1 XP_016870827.1 6, 7 743
          XM_017015339.1 XP_016870828.1 4, 5 671
          XM_017015340.1 XP_016870829.1 2, 3 191
          GenBank mRNA AB033084.1 - 4, 5 542
          AF019638.1 - 4, 5 551
          AF095286.1 - 4, 5 559
          AK097540.1 - 5, 6 774
          AK300418.1 - 4, 5 582
          AK315988.1 - 5, 6 494
          BC053584.1 - 4, 5 649 5186337

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          deaminase

          Gene Ontology Categories:

          Function(s) Process(es)

          nucleobase-containing compound metabolic process
          guanine catabolic process
          purine nucleotide catabolic process
          nervous system development
          guanine metabolic process
          zinc ion binding
          guanine deaminase activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-8jdkq:80/100.66.75.107:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0