Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SH3PXD2A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014631.2 | NP_055446.2 | 14 | 3405 | ||
XM_005270294.4 | XP_005270351.1 | 15 | 3538 | |||
XM_005270295.4 | XP_005270352.1 | 14 | 3492 | |||
XM_005270297.3 | XP_005270354.1 | 9 | 4769 | |||
XM_005270298.4 | XP_005270355.1 | 9 | 3572 | |||
XM_006718079.3 | XP_006718142.1 | 16 | 3635 | |||
XM_006718080.3 | XP_006718143.1 | 10 | 3666 | |||
XM_011540385.2 | XP_011538687.1 | 15 | 3589 | |||
XM_011540386.2 | XP_011538688.1 | 10 | 4865 | |||
XM_011540387.2 | XP_011538689.1 | 6 | 6511 | |||
XM_017016969.1 | XP_016872458.1 | 15 | 3556 | |||
XM_017016970.1 | XP_016872459.1 | 14 | 3372 | |||
GenBank mRNA | AB007878.1 | - | 12 | 3155 | ||
AK056469.1 | - | 13 | 3128 | |||
AK294454.1 | - | 9 | 2945 | |||
AK294736.1 | - | 9 | 2983 | |||
AK295660.1 | - | 9 | 3049 | |||
AK302015.1 | - | 13 | 2912 | |||
AK304828.1 | - | 9 | 2961 | |||
AK315983.1 | - | 9 | 2988 |