Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other HDAC4 SIRNA Assays ›
Gene Symbol:
HDAC4
Gene Name:
histone deacetylase 4
Gene Aliases:
AHO3, BDMR, HA6116, HD4, HDAC-4, HDAC-A, HDACA
Chromosome Location:
Chr.2: 239048168 - 239401647 on Build GRCh38
UniGene ID:
Hs.20516
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
HSS114674
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: HDAC4
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_006037.3
NP_006028.2
15
2769
XM_006712877.3
XP_006712940.1
14
8715
XM_006712878.3
XP_006712941.1
15
2289
XM_006712879.3
XP_006712942.1
14
8182
XM_006712880.3
XP_006712943.1
14
8194
XM_011512217.2
XP_011510519.1
14
8730
XM_011512218.2
XP_011510520.1
14
8730
XM_011512219.2
XP_011510521.1
14
8715
XM_011512220.2
XP_011510522.1
14
2469
XM_011512221.1
XP_011510523.1
15
2457
XM_011512222.2
XP_011510524.1
15
2758
XM_011512223.2
XP_011510525.1
15
2272
XM_011512224.2
XP_011510526.1
14
2660
XM_011512225.2
XP_011510527.1
14
2645
XM_011512226.2
XP_011510528.1
14
2217
XM_011512227.2
XP_011510529.1
13
7744
XM_011512230.1
XP_011510532.1
9
1655
XM_017005394.1
XP_016860883.1
13
8599
XM_017005395.1
XP_016860884.1
11
1521
GenBank mRNA
AB006626.2
-
15
2769
AF132607.1
-
14
1977
AK302719.1
-
9
1338
AK303368.1
-
14
2336
AK303467.1
-
13
1976
AK303617.1
-
12
1950
AK308791.1
-
15
2462
AK310603.1
-
14
2429
BC039904.1
-
15
2036
6067363
Back To Top
More Information
Panther Classification:
Molecular Function -
histone modifying enzyme
Gene Ontology Categories:
Function(s)
Process(es)
negative regulation of transcription from RNA polymerase II promoter
skeletal system development
osteoblast development
chromatin remodeling
transcription, DNA-templated
inflammatory response
nervous system development
positive regulation of cell proliferation
negative regulation of cell proliferation
positive regulation of lamellipodium assembly
negative regulation of myotube differentiation
regulation of cardiac muscle contraction by calcium ion signaling
response to denervation involved in regulation of muscle adaptation
cardiac muscle hypertrophy in response to stress
positive regulation of smooth muscle cell migration
chromatin modification
histone deacetylation
B cell differentiation
positive regulation of protein sumoylation
peptidyl-lysine deacetylation
regulation of gene expression, epigenetic
B cell activation
response to drug
regulation of protein binding
negative regulation of sequence-specific DNA binding transcription factor activity
positive regulation of neuron apoptotic process
negative regulation of osteoblast differentiation
negative regulation of glycolytic process
negative regulation of transcription, DNA-templated
positive regulation of transcription, DNA-templated
positive regulation of transcription from RNA polymerase II promoter
positive regulation of smooth muscle cell proliferation
regulation of skeletal muscle fiber development
positive regulation of sequence-specific DNA binding transcription factor activity
response to interleukin-1
histone H3 deacetylation
histone H4 deacetylation
cellular response to mechanical stimulus
cellular response to tumor necrosis factor
cellular response to parathyroid hormone stimulus
positive regulation of reactive oxygen species biosynthetic process
RNA polymerase III transcription factor binding
core promoter binding
chromatin binding
transcription corepressor activity
histone deacetylase activity
protein binding
transcription factor binding
zinc ion binding
protein kinase binding
potassium ion binding
NAD-dependent histone deacetylase activity (H3-K14 specific)
protein deacetylase activity
activating transcription factor binding
histone deacetylase binding
sequence-specific DNA binding
transcription regulatory region DNA binding
repressing transcription factor binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-659f68c6f4-zjghb:80/100.66.79.197:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0