Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MLEC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001303627.1 | NP_001290556.1 | 3 | 353 | ||
NM_014730.3 | NP_055545.1 | 3 | 636 | |||
XM_011539031.1 | XP_011537333.1 | 3 | 5051 | |||
XM_011539032.1 | XP_011537334.1 | 4 | 4739 | |||
GenBank mRNA | BC000371.2 | - | 3 | 585 | 2819615 | |
BC016297.2 | - | 3 | 587 | 3832682 | ||
BI458950.1 | - | 3 | 355 | |||
BU845446.1 | - | 614 | ||||
CA423273.1 | - | 3 | 572 | |||
D63486.1 | - | 3 | 591 |