Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
CDMO and Clinical Trial Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other ATG13 SIRNA Assays ›
Gene Symbol:
ATG13
Gene Name:
autophagy related 13
Gene Aliases:
KIAA0652, PARATARG8
Chromosome Location:
Chr.11: 46617276 - 46676019 on Build GRCh38
UniGene ID:
Hs.127403
Species:
Human
siRNA ID
HSS114712
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: ATG13
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001142673.2
NP_001136145.1
14
1780
NM_001205119.1
NP_001192048.1
14
1823
NM_001205120.1
NP_001192049.1
13
1724
NM_001205121.1
NP_001192050.1
13
1664
NM_001205122.1
NP_001192051.1
11
1531
NM_014741.4
NP_055556.2
13
1669
XM_005253262.2
XP_005253319.1
15
1619
XM_005253263.2
XP_005253320.1
15
1910
XM_005253265.2
XP_005253322.1
16
2116
XM_005253266.2
XP_005253323.1
15
1845
XM_005253268.2
XP_005253325.1
13
1222
XM_006718394.2
XP_006718457.1
16
1966
XM_006718395.2
XP_006718458.1
16
1901
XM_006718396.2
XP_006718459.1
15
2000
XM_011520493.1
XP_011518795.1
17
2103
XM_011520496.2
XP_011518798.1
14
1878
XM_011520499.2
XP_011518801.1
14
1492
XM_011520500.1
XP_011518802.1
16
1939
XM_011520501.1
XP_011518803.1
15
1867
XM_011520502.1
XP_011518804.1
15
1840
XM_011520503.1
XP_011518805.1
14
1756
XM_017018599.1
XP_016874088.1
16
2051
XM_017018600.1
XP_016874089.1
15
1592
XM_017018601.1
XP_016874090.1
14
1767
XM_017018602.1
XP_016874091.1
15
2017
XM_017018603.1
XP_016874092.1
16
2134
XM_017018604.1
XP_016874093.1
14
1811
XM_017018605.1
XP_016874094.1
15
1802
XM_017018606.1
XP_016874095.1
14
1746
XM_017018607.1
XP_016874096.1
12
1123
XM_017018608.1
XP_016874097.1
14
1493
XM_017018609.1
XP_016874098.1
14
1784
XM_017018610.1
XP_016874099.1
13
1668
XM_017018611.1
XP_016874100.1
14
1719
XM_017018612.1
XP_016874101.1
14
1906
XM_017018613.1
XP_016874102.1
13
1700
XM_017018614.1
XP_016874103.1
14
1691
XM_017018615.1
XP_016874104.1
13
1790
XM_017018616.1
XP_016874105.1
12
1584
XM_017018617.1
XP_016874106.1
14
1789
XM_017018618.1
XP_016874107.1
16
2105
XM_017018619.1
XP_016874108.1
14
1890
XM_017018620.1
XP_016874109.1
14
1950
XM_017018621.1
XP_016874110.1
13
1834
XM_017018622.1
XP_016874111.1
13
1791
GenBank mRNA
AB014552.1
-
13
1404
AK023867.1
-
13
1402
AK223603.1
-
14
1499
AK289641.1
-
14
1327
AK293109.1
-
13
1368
AK294110.1
-
11
1244
AK296526.1
-
6
611
AK315925.1
-
6
538
BC001331.1
-
13
1098
3458359
BC002378.2
-
13
1368
2961127
BC006191.2
-
13
1410
3936851
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
autophagosome assembly
macroautophagy
mitophagy in response to mitochondrial depolarization
positive regulation of protein targeting to mitochondrion
protein binding
protein kinase binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-75dc5564b9-9fjjm:80/10.239.165.64:80.
git-commit: 1b386f5412c20333c1bb415eea69346c63d8df9e
git-url: git@github.com:thermofisher/magellan-core.git
git-branch: origin/release/2.7.0-2024.04.17-1.0