Search
Search

EFCAB14| Gene Symbol: EFCAB14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_014774.2 | NP_055589.1 | 4 | 1540 | ||
| GenBank mRNA | AB007963.1 | - | 4 | 1540 | ||
| AK001487.1 | - | 1 | 39 | |||
| AK295722.1 | - | 2 | 182 | |||
| BC002525.2 | - | 4 | 685 | 3139273 | ||
| BC018842.2 | - | 4 | 685 | |||