Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Clinical Genomics
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other C2CD5 SIRNA Assays ›
Gene Symbol:
C2CD5
Gene Name:
C2 calcium dependent domain containing 5
Gene Aliases:
CDP138, KIAA0528
Chromosome Location:
Chr.12: 22448546 - 22544546 on Build GRCh38
UniGene ID:
Hs.271014
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
HSS114870
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Contact Us
Your Price
Online offer:
3062.0
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: C2CD5
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001286173.1
NP_001273102.1
27
3203
NM_001286174.1
NP_001273103.1
25
3337
NM_001286175.1
NP_001273104.1
27
3206
NM_001286176.1
NP_001273105.1
26
3197
NM_001286177.1
NP_001273106.1
26
3170
NM_014802.2
NP_055617.1
24
3044
XM_005253538.2
XP_005253595.1
24
2979
XM_005253539.3
XP_005253596.1
18
1889
XM_006719170.2
XP_006719233.1
25
3057
XM_017020264.1
XP_016875753.1
30
3489
XM_017020265.1
XP_016875754.1
30
3459
XM_017020266.1
XP_016875755.1
29
3438
XM_017020267.1
XP_016875756.1
28
3417
XM_017020268.1
XP_016875757.1
27
3351
XM_017020269.1
XP_016875758.1
28
3336
XM_017020270.1
XP_016875759.1
29
3297
XM_017020271.1
XP_016875760.1
29
3270
XM_017020272.1
XP_016875761.1
27
3198
XM_017020273.1
XP_016875762.1
26
3147
XM_017020274.1
XP_016875763.1
26
3129
XM_017020275.1
XP_016875764.1
27
3117
XM_017020276.1
XP_016875765.1
26
3102
XM_017020277.1
XP_016875766.1
25
3081
XM_017020278.1
XP_016875767.1
25
3045
XM_017020279.1
XP_016875768.1
22
2212
GenBank mRNA
AB011100.3
-
23
2797
AK295862.1
-
27
3151
AK299353.1
-
25
3044
AY166851.1
-
24
3016
BC042498.1
-
23
2803
5174801
BC053885.1
-
19
2268
BC117143.1
-
26
3033
40125694
BC143859.1
-
26
3060
9052375
BC143860.1
-
26
3033
9052376
BC143878.1
-
27
3066
9052394
BP418369.1
-
2
157
BX648560.1
-
7
661
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
vesicle fusion
positive regulation of glucose transport
positive regulation of vesicle fusion
cellular response to insulin stimulus
insulin receptor signaling pathway via phosphatidylinositol 3-kinase
intracellular protein transmembrane transport
protein localization to plasma membrane
positive regulation of protein targeting to membrane
positive regulation of glucose import in response to insulin stimulus
calcium ion binding
calcium-dependent phospholipid binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-55658555d6-pnd5g:80/100.66.79.83:80.
git-commit: ec8e7df5f8fec8d765bd419205f1b5046a016d37
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.41.0-Offline