Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other MED24 SIRNA Assays ›
          Gene Symbol:
          MED24
          Gene Name:
          mediator complex subunit 24
          Gene Aliases:
          ARC100, CRSP100, CRSP4, DRIP100, MED5, THRAP4, TRAP100
          Chromosome Location:
          Chr.17: 40019097 - 40054636 on Build GRCh38
          UniGene ID:
          Hs.462983
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID HSS114902
          Size 20 nmol
          Availability Made To Order
          Catalog # 1299001
          Price Contact Us
          Your Price
          Online offer:
          3062.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: MED24
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001079518.1 NP_001072986.1 18, 19 2287
          NM_001267797.1 NP_001254726.1 18, 19 2265
          NM_014815.3 NP_055630.2 19, 20 2304
          NR_052017.1 18, 19 2230
          XM_005257874.2 XP_005257931.1 19, 20 2209
          XM_006722204.2 XP_006722267.1 19, 20 2245
          XM_006722206.2 XP_006722269.1 19, 20 2153
          XM_006722207.2 XP_006722270.1 19, 20 2121
          XM_011525529.2 XP_011523831.1 20, 21 2302
          XM_011525530.2 XP_011523832.1 20, 21 2142
          XM_011525531.2 XP_011523833.1 19, 20 2085
          XM_011525532.2 XP_011523834.1 20, 21 2210
          XM_011525533.2 XP_011523835.1 20, 21 2178
          XM_011525534.2 XP_011523836.1 19, 20 2121
          XM_011525535.2 XP_011523837.1 19, 20 2121
          XM_011525536.2 XP_011523838.1 20, 21 2142
          XM_011525538.2 XP_011523840.1 19, 20 2436
          XM_017025458.1 XP_016880947.1 20, 21 2174
          XM_017025459.1 XP_016880948.1 19, 20 2117
          XM_017025460.1 XP_016880949.1 20, 21 2174
          XM_017025461.1 XP_016880950.1 20, 21 2266
          XM_017025462.1 XP_016880951.1 20, 21 2174
          XM_017025463.1 XP_016880952.1 20, 21 2142
          XM_017025464.1 XP_016880953.1 19, 20 2121
          XM_017025465.1 XP_016880954.1 19, 20 2245
          XM_017025466.1 XP_016880955.1 19, 20 2117
          XM_017025467.1 XP_016880956.1 19, 20 2117
          XM_017025468.1 XP_016880957.1 19, 20 2085
          XM_017025469.1 XP_016880958.1 19, 20 2400
          XM_017025470.1 XP_016880959.1 19, 20 2400
          GenBank mRNA AF055995.1 - 18, 19 1991
          AF277379.1 - 18, 19 1991
          AK022508.1 - 18, 19 2056
          AK291040.1 - 18, 19 2036
          AK293309.1 - 17, 18 1862
          AK299864.1 - 19, 20 1921
          AK300992.1 - 18, 19 1909
          AK303502.1 - 7, 8 759
          AK303745.1 - 18, 19 2000
          BC011375.1 - 18, 19 2266 3866038
          D50920.1 - 19, 20 2064
          EU392542.1 - 4, 5 349

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          general transcription factor

          Gene Ontology Categories:

          Function(s) Process(es)

          regulation of transcription from RNA polymerase II promoter
          transcription initiation from RNA polymerase II promoter
          protein ubiquitination
          histone acetylation
          stem cell population maintenance
          intracellular steroid hormone receptor signaling pathway
          androgen receptor signaling pathway
          positive regulation of transcription, DNA-templated
          protein heterooligomerization
          RNA polymerase II transcription cofactor activity
          transcription cofactor activity
          histone acetyltransferase activity
          receptor activity
          protein binding
          ligand-dependent nuclear receptor transcription coactivator activity
          vitamin D receptor binding
          thyroid hormone receptor binding
          ubiquitin protein ligase activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-wf7gw:80/100.66.73.223:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0