Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other SNAP91 SIRNA Assays ›
Gene Symbol:
SNAP91
Gene Name:
synaptosome associated protein 91
Gene Aliases:
AP180, CALM
Chromosome Location:
Chr.6: 83552885 - 83709408 on Build GRCh38
UniGene ID:
Hs.368046
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
HSS114952
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: SNAP91
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001242792.1
NP_001229721.1
21
2211
NM_001256718.1
NP_001243647.1
20
1921
NM_014841.2
NP_055656.1
21
2214
XM_005248770.4
XP_005248827.1
21
2053
XM_006715615.1
XP_006715678.1
21
2005
XM_011536265.1
XP_011534567.1
21
2012
XM_011536266.1
XP_011534568.1
21
2124
XM_011536269.1
XP_011534571.1
21
2005
XM_011536271.1
XP_011534573.1
20
1963
XM_011536273.2
XP_011534575.1
20
1927
XM_011536276.2
XP_011534578.1
20
1897
XM_011536277.1
XP_011534579.1
23
2293
XM_017011554.1
XP_016867043.1
20
2163
XM_017011555.1
XP_016867044.1
20
2047
XM_017011556.1
XP_016867045.1
20
1979
XM_017011557.1
XP_016867046.1
21
2182
XM_017011558.1
XP_016867047.1
21
2012
XM_017011559.1
XP_016867048.1
21
2111
XM_017011560.1
XP_016867049.1
21
2053
XM_017011561.1
XP_016867050.1
21
2124
XM_017011562.1
XP_016867051.1
20
2167
XM_017011563.1
XP_016867052.1
19
2161
XM_017011564.1
XP_016867053.1
19
2032
XM_017011565.1
XP_016867054.1
19
2090
XM_017011566.1
XP_016867055.1
20
2140
XM_017011567.1
XP_016867056.1
20
1943
XM_017011568.1
XP_016867057.1
19
2134
XM_017011569.1
XP_016867058.1
20
2140
XM_017011570.1
XP_016867059.1
20
2104
XM_017011571.1
XP_016867060.1
20
1975
XM_017011572.1
XP_016867061.1
20
1934
XM_017011573.1
XP_016867062.1
20
2033
XM_017011574.1
XP_016867063.1
21
2182
XM_017011580.1
XP_016867069.1
20
2074
XM_017011581.1
XP_016867070.1
20
1904
XM_017011582.1
XP_016867071.1
20
1933
XM_017011583.1
XP_016867072.1
20
2061
XM_017011584.1
XP_016867073.1
20
1904
XM_017011585.1
XP_016867074.1
20
1945
XM_017011586.1
XP_016867075.1
20
1897
XM_017011587.1
XP_016867076.1
20
2003
XM_017011588.1
XP_016867077.1
20
2016
XM_017011589.1
XP_016867078.1
19
2032
GenBank mRNA
AB014556.1
-
21
2140
AK289582.1
-
21
2182
AK294556.1
-
14
1172
AK294898.1
-
20
1905
Back To Top
More Information
Panther Classification:
Molecular Function -
vesicle coat protein
Gene Ontology Categories:
Function(s)
Process(es)
chemical synaptic transmission
protein transport
clathrin coat assembly
regulation of clathrin-mediated endocytosis
protein binding
1-phosphatidylinositol binding
protein kinase binding
clathrin binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-659f68c6f4-wf7gw:80/100.66.73.223:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0