Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Cell Analysis
Antibodies
Mass Spectrometry
Cell Culture
Laboratory Instruments
Clinical and Diagnostics
Chromatography
Laboratory Equipment
Laboratory Supplies
Molecular Biology and Nucleic Acid Analysis
Sequence-Specific Nucleic Acid Products
See all product categories
Applications
Cell Culture and Transfection
Flow Cytometry
Cancer Research
Chromatography
Sequencing
PCR
Lab Solutions
Allergy Diagnostics
See all applications and techniques
Services
Cell Biology Services
Custom Services
Training Services
Lab Informatics Services
Financial and Leasing Services
Partnering and Licensing Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Food Safety Inspection Services
See all services
Help and Support
How to Order
Promotions and Online Offers
Contact Us
Change Location
Create a New Account
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Special Offers
Contact Us
Quick Order
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Contact Us
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other NR1I3 SIRNA Assays ›
Gene Symbol:
NR1I3
Gene Name:
nuclear receptor subfamily 1 group I member 3
Gene Aliases:
CAR, CAR1, MB67
Chromosome Location:
Chr.1: 161229666 - 161238623 on Build GRCh38
UniGene ID:
Hs.349642
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
HSS115121
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: NR1I3
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001077469.2
NP_001070937.1
4
590
NM_001077470.2
NP_001070938.1
3
450
NM_001077471.2
NP_001070939.1
4
590
NM_001077472.2
NP_001070940.1
3
450
NM_001077473.2
NP_001070941.1
3
450
NM_001077474.2
NP_001070942.1
4
590
NM_001077475.2
NP_001070943.1
3
450
NM_001077476.2
NP_001070944.1
3
450
NM_001077477.2
NP_001070945.1
3
450
NM_001077478.2
NP_001070946.1
4
590
NM_001077479.2
NP_001070947.1
3
450
NM_001077480.2
NP_001070948.1
4
590
NM_001077481.2
NP_001070949.1
4
590
NM_001077482.2
NP_001070950.1
4
590
NM_005122.4
NP_005113.1
4
590
XM_005245693.4
XP_005245750.1
3
863
XM_005245694.4
XP_005245751.1
3
862
XM_005245697.4
XP_005245754.1
4
473
XM_011510237.2
XP_011508539.1
3
860
GenBank mRNA
AB307702.1
-
3
387
AK303757.1
-
3
443
AK303796.1
-
4
624
AK316208.1
-
4
582
AY572806.1
-
4
481
AY572807.1
-
4
410
AY572808.1
-
3
341
AY572810.1
-
4
481
AY572811.1
-
3
341
AY572812.1
-
3
341
AY572813.1
-
4
481
AY572816.1
-
3
341
AY572817.1
-
3
341
AY572818.1
-
3
341
AY572819.1
-
4
481
AY572827.1
-
3
341
BC069626.1
-
4
543
7262456
BC121120.1
-
4
491
40120664
BC121121.1
-
4
491
40120665
CB124867.1
-
4
590
DQ022681.1
-
3
387
HQ692838.1
-
3
387
HQ692839.1
-
3
387
HQ692840.1
-
3
387
HQ692841.1
-
3
387
HQ709178.1
-
3
387
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
osteoblast differentiation
transcription initiation from RNA polymerase II promoter
signal transduction
androgen receptor signaling pathway
negative regulation of transcription, DNA-templated
positive regulation of transcription from RNA polymerase II promoter
RNA polymerase II regulatory region sequence-specific DNA binding
transcriptional activator activity, RNA polymerase II transcription regulatory region sequence-specific binding
DNA binding
transcription factor activity, sequence-specific DNA binding
transcription coactivator activity
RNA polymerase II transcription factor activity, ligand-activated sequence-specific DNA binding
androgen receptor activity
thyroid hormone receptor activity
zinc ion binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-xmxnb:80/100.66.76.46:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0