Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other NR1H4 SIRNA Assays ›
          Gene Symbol:
          NR1H4
          Gene Name:
          nuclear receptor subfamily 1 group H member 4
          Gene Aliases:
          BAR, FXR, HRR-1, HRR1, RIP14
          Chromosome Location:
          Chr.12: 100473773 - 100563867 on Build GRCh38
          UniGene ID:
          Hs.282735
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID HSS115123
          Size 20 nmol
          Availability Made To Order
          Catalog # 1299001
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: NR1H4
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001206977.1 NP_001193906.1 10, 11 1717
          NM_001206978.1 NP_001193907.1 8, 9 1397
          NM_001206979.1 NP_001193908.1 9, 10 1550
          NM_001206992.1 NP_001193921.1 7, 8 1120
          NM_001206993.1 NP_001193922.1 7, 8 1132
          NM_005123.3 NP_005114.1 9, 10 1538
          NR_135146.1 9, 10 1543
          XM_011539040.2 XP_011537342.1 9, 10 2468
          XM_011539041.2 XP_011537343.1 6, 7 1377
          GenBank mRNA AB307699.1 - 7, 8 1062
          AF384555.1 - 7, 8 1074
          AF478445.1 - 7, 8 1120
          AF478446.1 - 7, 8 1132
          AK296612.1 - 7, 8 1133
          AK296647.1 - 7, 8 1068
          AK312959.1 - 9, 10 1539
          BC071778.1 - 7, 8 1072
          BC130573.1 - 9, 10 1407 40146604
          BC144183.1 - 8, 9 1255
          BC144184.1 - 9, 10 1407 9052704
          BX571749.1 - 7, 8 1117
          HQ709175.1 - 7, 8 1074
          U68233.1 - 9, 10 1415

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of transcription from RNA polymerase II promoter
          nitrogen catabolite activation of transcription from RNA polymerase II promoter
          regulation of carbohydrate metabolic process
          transcription initiation from RNA polymerase II promoter
          inflammatory response
          signal transduction
          Notch signaling pathway
          bile acid metabolic process
          response to glucose
          regulation of low-density lipoprotein particle clearance
          bile acid and bile salt transport
          intracellular receptor signaling pathway
          response to lipopolysaccharide
          toll-like receptor 4 signaling pathway
          regulation of urea metabolic process
          histone H3-R17 methylation
          cellular triglyceride homeostasis
          bile acid signaling pathway
          intracellular bile acid receptor signaling pathway
          negative regulation of apoptotic process
          negative regulation of I-kappaB kinase/NF-kappaB signaling
          steroid hormone mediated signaling pathway
          innate immune response
          positive regulation of transcription from RNA polymerase II promoter
          digestive tract development
          regulation of insulin secretion involved in cellular response to glucose stimulus
          regulation of bile acid biosynthetic process
          negative regulation of bile acid biosynthetic process
          cellular response to fatty acid
          cellular response to organonitrogen compound
          interleukin-17 secretion
          regulation of cholesterol metabolic process
          negative regulation of interferon-gamma secretion
          negative regulation of tumor necrosis factor secretion
          positive regulation of glutamate metabolic process
          positive regulation of ammonia assimilation cycle
          RNA polymerase II distal enhancer sequence-specific DNA binding
          transcriptional activator activity, RNA polymerase II core promoter proximal region sequence-specific binding
          transcriptional activator activity, RNA polymerase II transcription factor binding
          transcription factor activity, sequence-specific DNA binding
          transcription factor activity, RNA polymerase II distal enhancer sequence-specific binding
          steroid hormone receptor activity
          transcription coactivator activity
          transcription corepressor activity
          RNA polymerase II transcription factor activity, ligand-activated sequence-specific DNA binding
          thyroid hormone receptor activity
          protein binding
          zinc ion binding
          ligand-dependent nuclear receptor binding
          bile acid binding
          bile acid receptor activity
          peptide binding
          sequence-specific DNA binding
          retinoid X receptor binding
          chenodeoxycholic acid binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-4sktr:80/100.66.76.202:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0