Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HNRNPDL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001207000.1 | NP_001193929.1 | 2 | 1034 | ||
NM_031372.3 | NP_112740.1 | 2 | 1034 | |||
NR_003249.2 | 2 | 1034 | ||||
GenBank mRNA | AB017019.1 | - | 2 | 1034 | ||
AB066484.1 | - | 2 | 142 | |||
AK300122.1 | - | 2 | 120 | |||
AY453824.1 | - | 2 | 133 | |||
BC007392.2 | - | 2 | 486 | |||
BC011714.2 | - | 2 | 675 | 3929324 | ||
BC018961.2 | - | 2 | 675 | |||
BC071944.1 | - | 2 | 615 | 5015862 | ||
CR407623.1 | - | 2 | 499 | |||
D89092.1 | - | 2 | 250 | |||
D89678.1 | - | 2 | 109 |