Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
CDMO and Clinical Trial Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other FAM13A SIRNA Assays ›
Gene Symbol:
FAM13A
Gene Name:
family with sequence similarity 13 member A
Gene Aliases:
ARHGAP48, FAM13A1
Chromosome Location:
Chr.4: 88725954 - 89111398 on Build GRCh38
UniGene ID:
Hs.714536
Species:
Human
siRNA ID
HSS115442
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: FAM13A
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001015045.2
NP_001015045.1
10
1392
NM_001265578.1
NP_001252507.1
9
1350
NM_001265579.1
NP_001252508.1
10
1392
NM_001265580.1
NP_001252509.1
8
1308
NM_014883.3
NP_055698.2
16
2244
XM_005262681.2
XP_005262738.1
15
2202
XM_005262682.2
XP_005262739.1
16
2343
XM_005262683.2
XP_005262740.1
16
2244
XM_005262684.3
XP_005262741.1
15
2023
XM_006714057.3
XP_006714120.1
13
1519
XM_011531516.1
XP_011529818.1
17
2265
XM_011531517.1
XP_011529819.1
15
2160
XM_011531518.1
XP_011529820.1
17
2394
XM_011531519.2
XP_011529821.1
15
1953
XM_017007624.1
XP_016863113.1
15
2202
XM_017007625.1
XP_016863114.1
15
1863
XM_017007626.1
XP_016863115.1
13
1497
XM_017007627.1
XP_016863116.1
15
2088
XM_017007628.1
XP_016863117.1
14
1981
XM_017007629.1
XP_016863118.1
16
3975
XM_017007630.1
XP_016863119.1
14
1888
XM_017007631.1
XP_016863120.1
15
3933
XM_017007632.1
XP_016863121.1
13
1827
XM_017007633.1
XP_016863122.1
15
3891
XM_017007634.1
XP_016863123.1
9
1301
GenBank mRNA
AB020721.2
-
10
1109
AK027138.1
-
10
1232
AK091328.1
-
3
1003
AK091787.1
-
9
1510
AK296932.1
-
8
1177
BC086875.1
-
9
1191
30708091
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
signal transduction
positive regulation of GTPase activity
regulation of small GTPase mediated signal transduction
GTPase activator activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-5c7f9f6c68-tvqvm:80/10.239.167.222:80.
git-commit: b16971a8df2afcd8df684aafbc9bf1e9f45e42d4
git-url: git@github.com:thermofisher/magellan-core.git
git-branch: origin/release/2.6.0-2024.04.15-1.0