Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RBM6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001167582.1 | NP_001161054.1 | 8, 9 | 998 | ||
NM_005777.2 | NP_005768.1 | 12, 13 | 2511 | |||
XM_005264784.1 | XP_005264841.1 | 12, 13 | 2786 | |||
XM_005264785.1 | XP_005264842.1 | 11, 12 | 1202 | |||
XM_005264786.1 | XP_005264843.1 | 10, 11 | 1100 | |||
XM_005264787.2 | XP_005264844.1 | 9, 10 | 1866 | |||
XM_006712916.1 | XP_006712979.1 | 12, 13 | 2611 | |||
XM_017005497.1 | XP_016860986.1 | 8, 9 | 835 | |||
XM_017005498.1 | XP_016860987.1 | 9, 10 | 1024 | |||
XM_017005499.1 | XP_016860988.1 | 14, 15 | 3071 | |||
XM_017005500.1 | XP_016860989.1 | 13, 14 | 2953 | |||
XM_017005501.1 | XP_016860990.1 | 13, 14 | 2904 | |||
XM_017005502.1 | XP_016860991.1 | 8, 9 | 899 | |||
GenBank mRNA | AF042857.1 | - | 12, 13 | 2364 | ||
AF069517.1 | - | 12, 13 | 2467 | |||
AF091264.1 | - | 12, 13 | 2396 | |||
AK093836.1 | - | 10, 11 | 1043 | |||
AK291719.1 | - | 12, 13 | 2511 | |||
AK298108.1 | - | 7, 8 | 904 | |||
AK308794.1 | - | 11, 12 | 1126 | |||
AL832236.1 | - | 8, 9 | 946 | |||
BC046643.1 | - | 8, 9 | 852 | 5753635 | ||
BX648535.1 | - | 12, 13 | 2688 | |||
U50839.1 | - | 11, 12 | 2297 |