Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MPHOSPH9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022782.3 | NP_073619.3 | 11 | 2223 | ||
NR_103517.1 | 11 | 1962 | ||||
XM_011537740.2 | XP_011536042.1 | 11 | 2127 | |||
XM_011537741.2 | XP_011536043.1 | 11 | 2127 | |||
XM_017018673.1 | XP_016874162.1 | 11 | 2221 | |||
XM_017018674.1 | XP_016874163.1 | 11 | 2127 | |||
XM_017018675.1 | XP_016874164.1 | 10 | 2029 | |||
XM_017018677.1 | XP_016874166.1 | 11 | 2127 | |||
XM_017018678.1 | XP_016874167.1 | 10 | 1947 | |||
XM_017018679.1 | XP_016874168.1 | 12 | 2160 | |||
XM_017018680.1 | XP_016874169.1 | 10 | 2069 | |||
XM_017018681.1 | XP_016874170.1 | 10 | 1982 | |||
GenBank mRNA | AK023016.1 | - | 6 | 946 | ||
AK091181.1 | - | 7 | 1552 | |||
AK096541.1 | - | 11 | 1835 | |||
AL096751.1 | - | 5 | 883 | |||
BC130440.1 | - | 7 | 1505 | 40146471 | ||
BC144016.1 | - | 6 | 1379 | |||
BC144017.1 | - | 6 | 1157 |