Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: USPL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001321532.1 | NP_001308461.1 | 8 | 2558 | ||
NM_001321533.1 | NP_001308462.1 | 7 | 1918 | |||
NM_001321534.1 | NP_001308463.1 | 6 | 1647 | |||
NM_005800.4 | NP_005791.3 | 9 | 2711 | |||
XM_005266213.3 | XP_005266270.1 | 10 | 3069 | |||
XM_006719751.3 | XP_006719814.1 | 9 | 2989 | |||
XM_017020314.1 | XP_016875803.1 | 8 | 2893 | |||
XM_017020315.1 | XP_016875804.1 | 7 | 2264 | |||
XM_017020316.1 | XP_016875805.1 | 9 | 2902 | |||
GenBank mRNA | AK057514.1 | - | 6 | 1647 | ||
AK289826.1 | - | 9 | 2467 | |||
BC036027.1 | - | 9 | 2483 | 5271146 | ||
BC038103.2 | - | 9 | 2621 | 4821753 | ||
CR627370.1 | - | 7 | 1661 | |||
X59131.1 | - | 9 | 2456 |