Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
CDMO and Clinical Trial Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other HNRNPR SIRNA Assays ›
Gene Symbol:
HNRNPR
Gene Name:
heterogeneous nuclear ribonucleoprotein R
Gene Aliases:
HNRPR, hnRNP-R
Chromosome Location:
Chr.1: 23304688 - 23344650 on Build GRCh38
UniGene ID:
Hs.373763
Species:
Human
siRNA ID
HSS115623
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: HNRNPR
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001102397.2
NP_001095867.1
10
1881
NM_001102398.2
NP_001095868.1
11
2056
NM_001102399.2
NP_001095869.1
10
1890
NM_001297620.1
NP_001284549.1
10
1933
NM_001297621.1
NP_001284550.1
9
1704
NM_001297622.1
NP_001284551.1
9
1767
NM_005826.4
NP_005817.1
11
2047
XM_005245711.4
XP_005245768.1
11
1969
XM_011540471.2
XP_011538773.1
11
1978
XM_011540472.2
XP_011538774.1
9
1728
XM_011540474.2
XP_011538776.1
10
2231
XM_011540475.2
XP_011538777.1
10
2228
XM_011540476.2
XP_011538778.1
9
2062
XM_011540477.2
XP_011538779.1
9
1999
XM_017000008.1
XP_016855497.1
10
2222
XM_017000009.1
XP_016855498.1
10
1792
XM_017000010.1
XP_016855499.1
10
1807
GenBank mRNA
AF000364.1
-
11
1945
AK098580.1
-
3
949
AK297382.1
-
10
1850
AK297405.1
-
9
1622
AK300029.1
-
9
1700
AK313142.1
-
11
1995
BC001449.2
-
11
2002
3139052
BC110389.1
-
10
1808
BX538163.1
-
3
798
BX640912.1
-
10
1826
BX647784.1
-
10
1788
DQ351905.1
-
9
1770
Back To Top
More Information
Panther Classification:
Molecular Function -
RNA metabolism protein
Gene Ontology Categories:
Function(s)
Process(es)
mRNA splicing, via spliceosome
mRNA processing
circadian rhythm
gene expression
negative regulation of catalytic activity
positive regulation of mRNA catabolic process
mRNA destabilization
nucleotide binding
RNA binding
mRNA 3'-UTR binding
protein binding
poly(A) RNA binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-55874ff697-sdrtx:80/10.239.162.255:80.
git-commit: a2f5e26549390d543f5d2d1fbad8092df65fbda4
git-url: git@github.com:thermofisher/magellan-core.git
git-branch: origin/release/2.10.0-2024.06.23-1.0