Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RABEPK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001174152.1 | NP_001167623.1 | 9 | 1120 | ||
NM_001174153.1 | NP_001167624.1 | 8 | 967 | |||
NM_005833.3 | NP_005824.2 | 8 | 1281 | |||
XM_005251640.4 | XP_005251697.1 | 8 | 1193 | |||
XM_005251641.4 | XP_005251698.1 | 7 | 1048 | |||
XM_005251642.4 | XP_005251699.1 | 7 | 1039 | |||
XM_005251644.4 | XP_005251701.1 | 6 | 889 | |||
XM_011518120.2 | XP_011516422.1 | 8 | 1355 | |||
XM_017014177.1 | XP_016869666.1 | 9 | 1105 | |||
XM_017014178.1 | XP_016869667.1 | 8 | 1007 | |||
XM_017014179.1 | XP_016869668.1 | 7 | 1080 | |||
XM_017014180.1 | XP_016869669.1 | 7 | 854 | |||
XM_017014181.1 | XP_016869670.1 | 6 | 965 | |||
XM_017014182.1 | XP_016869671.1 | 7 | 790 | |||
XM_017014183.1 | XP_016869672.1 | 7 | 1286 | |||
GenBank mRNA | AK290768.1 | - | 8 | 1125 | ||
BC000503.2 | - | 8 | 1258 | 2821471 | ||
BC047023.1 | - | 8 | 947 | 5396980 | ||
BC053541.1 | - | 8 | 1090 | 5724458 | ||
BC065725.1 | - | 9 | 1123 | 4102355 | ||
CR456837.1 | - | 7 | 951 | |||
CR541851.1 | - | 7 | 951 | |||
KU178507.1 | - | 7 | 951 | |||
KU178508.1 | - | 7 | 864 | |||
KU178509.1 | - | 6 | 802 | |||
KU178510.1 | - | 6 | 798 | |||
KU178511.1 | - | 5 | 636 | |||
Z97074.1 | - | 9 | 1100 |