Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NET1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001047160.2 | NP_001040625.1 | 12 | 1835 | ||
NM_005863.4 | NP_005854.2 | 10 | 1746 | |||
NR_073040.1 | 10 | 1779 | ||||
GenBank mRNA | AB209852.1 | - | 12 | 1781 | ||
AJ010046.1 | - | 10 | 1679 | |||
AK024919.1 | - | 10 | 1686 | |||
AK294244.1 | - | 6 | 1212 | |||
AK299335.1 | - | 12 | 1791 | |||
AK302687.1 | - | 12 | 1617 | |||
AK304028.1 | - | 10 | 1721 | |||
AK315566.1 | - | 10 | 1688 | |||
BC010285.1 | - | 10 | 1677 | 3048980 | ||
BC053553.1 | - | 12 | 1787 | 6137683 | ||
BX537509.1 | - | 6 | 4384 | |||
U02081.1 | - | 8 | 1267 |