Search
Search

APBB3| Gene Symbol: APBB3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_006051.3 | NP_006042.3 | 10 | 1252 | ||
| NM_133172.2 | NP_573418.2 | 9 | 1246 | |||
| NM_133173.2 | NP_573419.2 | 10 | 1231 | |||
| NM_133174.2 | NP_573420.2 | 9 | 1225 | |||
| GenBank mRNA | AB018247.1 | - | 10 | 923 | ||
| AB049618.1 | - | 10 | 1115 | |||
| AB061224.1 | - | 10 | 1018 | |||
| AB079687.1 | - | 10 | 872 | |||
| AB094990.1 | - | 9 | 866 | |||
| AB209839.1 | - | 10 | 959 | |||
| AF224708.1 | - | 9 | 907 | |||
| AF224709.1 | - | 10 | 892 | |||
| AF224710.1 | - | 9 | 886 | |||
| AF224711.1 | - | 10 | 913 | |||
| AK074885.1 | - | 5 | 1158 | |||
| AK075280.1 | - | 10 | 1194 | |||
| AK125244.1 | - | 7 | 1884 | |||
| AK304824.1 | - | 6 | 1252 | |||
| BC013158.1 | - | 10 | 1231 | 3870148 | ||
| BC125187.1 | - | 10 | 1223 | 40122344 | ||