Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PFDN6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001185181.2 | NP_001172110.1 | 3, 4 | 482 | ||
NM_001265595.1 | NP_001252524.1 | 4, 5 | 374 | |||
NM_001265596.1 | NP_001252525.1 | 4, 5 | 371 | |||
NM_014260.3 | NP_055075.1 | 4, 5 | 379 | |||
GenBank mRNA | AK312150.1 | - | 4, 5 | 367 | ||
BC039033.1 | - | 3, 4 | 348 | 5748799 | ||
BC059783.1 | - | 3, 4 | 354 | 5415221 | ||
BE794658.1 | - | 3, 4 | 335 | |||
BM686351.1 | - | 3, 4 | 463 | |||
BM745352.1 | - | 4, 5 | 374 | |||
BM792413.1 | - | 3, 4 | 356 | |||
BM818422.1 | - | 4, 5 | 374 | |||
BU734446.1 | - | 1, 2 | 232 | |||
DA256816.1 | - | 4, 5 | 367 |