Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other TACC2 SIRNA Assays ›
Gene Symbol:
TACC2
Gene Name:
transforming acidic coiled-coil containing protein 2
Gene Aliases:
AZU-1, ECTACC
Chromosome Location:
Chr.10: 121989174 - 122254545 on Build GRCh38
UniGene ID:
Hs.501252
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
HSS116287
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: TACC2
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001291876.1
NP_001278805.1
8
7226
NM_001291877.1
NP_001278806.1
8
7373
NM_001291878.1
NP_001278807.1
3
1734
NM_001291879.1
NP_001278808.1
5
1584
NM_006997.3
NP_008928.1
3
1734
NM_206860.2
NP_996742.1
3
1734
NM_206861.2
NP_996743.1
6
1799
NM_206862.3
NP_996744.3
9
7361
XM_005269388.1
XP_005269445.1
10
7185
XM_005269389.4
XP_005269446.1
10
7709
XM_005269390.1
XP_005269447.1
10
7185
XM_005269391.1
XP_005269448.1
9
7125
XM_005269392.1
XP_005269449.1
10
7185
XM_005269393.1
XP_005269450.1
10
7185
XM_005269394.1
XP_005269451.1
9
7059
XM_005269395.1
XP_005269452.1
9
7050
XM_005269396.1
XP_005269453.1
10
7185
XM_005269397.1
XP_005269454.1
8
6924
XM_005269399.1
XP_005269456.1
9
7050
XM_006717548.1
XP_006717611.1
10
7185
XM_006717552.1
XP_006717615.1
10
7185
XM_006717553.1
XP_006717616.1
10
7185
XM_006717554.1
XP_006717617.1
10
7185
XM_011539126.1
XP_011537428.1
10
7182
XM_011539128.1
XP_011537430.1
8
1758
XM_011539129.1
XP_011537431.1
7
1623
XM_011539130.1
XP_011537432.1
7
1623
XM_011539131.1
XP_011537433.1
3
1596
XM_011539132.1
XP_011537434.1
3
1596
XM_011539133.1
XP_011537435.1
4
1537
XM_011539134.1
XP_011537436.1
4
1808
XM_017015484.1
XP_016870973.1
10
7185
XM_017015485.1
XP_016870974.1
10
7185
XM_017015486.1
XP_016870975.1
10
7185
XM_017015487.1
XP_016870976.1
10
7185
XM_017015488.1
XP_016870977.1
8
6924
XM_017015489.1
XP_016870978.1
8
6924
XM_017015491.1
XP_016870980.1
3
1736
XM_017015492.1
XP_016870981.1
3
1736
XM_017015493.1
XP_016870982.1
3
1596
XM_017015494.1
XP_016870983.1
3
1736
XM_017015495.1
XP_016870984.1
3
1596
XM_017015496.1
XP_016870985.1
4
1537
XM_017015497.1
XP_016870986.1
4
1808
XM_017015498.1
XP_016870987.1
3
1673
XM_017015499.1
XP_016870988.1
5
1777
GenBank mRNA
AF095791.2
-
3
1321
AF176646.1
-
4
1722
AF220152.2
-
1
1398
AF528098.1
-
6
1780
AF528099.1
-
9
7342
AK027066.1
-
1
505
AK295631.1
-
3
1517
AL713712.1
-
1
810
AL833304.1
-
8
6460
BC039311.1
-
3
1734
BC141837.1
-
4
1386
BC142673.1
-
4
1386
BC144599.1
-
8
7341
9053129
BC144600.1
-
8
7194
9053130
BC150627.1
-
8
7194
9056997
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
microtubule cytoskeleton organization
cell proliferation
cerebral cortex development
nuclear hormone receptor binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-mtn5c:80/100.66.73.196:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0