Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
CDMO and Clinical Trial Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other SORBS1 SIRNA Assays ›
Gene Symbol:
SORBS1
Gene Name:
sorbin and SH3 domain containing 1
Gene Aliases:
CAP, FLAF2, R85FL, SH3D5, SH3P12, SORB1
Chromosome Location:
Chr.10: 95311773 - 95561420 on Build GRCh38
UniGene ID:
Hs.38621
Species:
Human
siRNA ID
HSS116291
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: SORBS1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001034954.1
NP_001030126.1
29, 30
3732
NM_001034955.1
NP_001030127.1
29, 30
3309
NM_001034956.1
NP_001030128.1
26, 27
2571
NM_001034957.1
NP_001030129.1
20, 21
2076
NM_001290294.1
NP_001277223.1
30, 31
3841
NM_001290295.1
NP_001277224.1
23, 24
2290
NM_001290296.1
NP_001277225.1
26, 27
2868
NM_001290297.1
NP_001277226.1
23, 24
2289
NM_001290298.1
NP_001277227.1
24, 25
2394
NM_006434.2
NP_006425.2
19, 20
1908
NM_015385.3
NP_056200.1
23, 24
2491
NM_024991.2
NP_079267.1
24, 25
2386
XM_006717589.1
XP_006717652.1
28, 29
3008
XM_006717593.1
XP_006717656.1
22, 23
2241
XM_011539140.1
XP_011537442.1
32, 33
4586
XM_011539150.1
XP_011537452.1
31, 32
4511
XM_011539155.1
XP_011537457.1
26, 27
4161
XM_011539167.1
XP_011537469.1
25, 26
3444
XM_017015500.1
XP_016870989.1
27, 28
4091
XM_017015501.1
XP_016870990.1
29, 30
3677
XM_017015502.1
XP_016870991.1
28, 29
3584
XM_017015503.1
XP_016870992.1
27, 28
3509
XM_017015504.1
XP_016870993.1
30, 31
3500
XM_017015505.1
XP_016870994.1
26, 27
3425
XM_017015506.1
XP_016870995.1
29, 30
3425
XM_017015507.1
XP_016870996.1
27, 28
3416
XM_017015508.1
XP_016870997.1
28, 29
3332
XM_017015509.1
XP_016870998.1
28, 29
3354
XM_017015510.1
XP_016870999.1
25, 26
3257
XM_017015511.1
XP_016871000.1
27, 28
3248
XM_017015512.1
XP_016871001.1
25, 26
3159
XM_017015513.1
XP_016871002.1
25, 26
3150
XM_017015514.1
XP_016871003.1
25, 26
3066
XM_017015515.1
XP_016871004.1
27, 28
2867
XM_017015516.1
XP_016871005.1
26, 27
2840
XM_017015517.1
XP_016871006.1
26, 27
2783
XM_017015518.1
XP_016871007.1
26, 27
2796
XM_017015519.1
XP_016871008.1
25, 26
2769
XM_017015520.1
XP_016871009.1
25, 26
2708
XM_017015521.1
XP_016871010.1
25, 26
2721
XM_017015522.1
XP_016871011.1
24, 25
2681
XM_017015523.1
XP_016871012.1
24, 25
2637
XM_017015524.1
XP_016871013.1
23, 24
2610
XM_017015525.1
XP_016871014.1
27, 28
2595
XM_017015526.1
XP_016871015.1
26, 27
2498
XM_017015527.1
XP_016871016.1
26, 27
2520
XM_017015528.1
XP_016871017.1
25, 26
2471
XM_017015529.1
XP_016871018.1
25, 26
2423
XM_017015530.1
XP_016871019.1
25, 26
2414
XM_017015531.1
XP_016871020.1
25, 26
2427
XM_017015532.1
XP_016871021.1
24, 25
2396
XM_017015533.1
XP_016871022.1
24, 25
2400
XM_017015534.1
XP_016871023.1
25, 26
2330
XM_017015535.1
XP_016871024.1
24, 25
2352
XM_017015536.1
XP_016871025.1
24, 25
2343
XM_017015537.1
XP_016871026.1
23, 24
2312
XM_017015538.1
XP_016871027.1
24, 25
2303
XM_017015539.1
XP_016871028.1
23, 24
2325
XM_017015540.1
XP_016871029.1
23, 24
2316
GenBank mRNA
AB037717.1
-
24, 25
2318
AF136380.1
-
18, 19
1863
AF136381.1
-
25, 26
2823
AF330623.1
-
22, 23
2244
AF330624.1
-
8, 9
942
AF356525.1
-
29, 30
3743
AF356526.1
-
29, 30
3320
AF356527.1
-
26, 27
2582
AJ489942.1
-
23, 24
2349
AK294924.1
-
12, 13
1268
AK300407.1
-
23, 24
2243
AK304130.1
-
12, 13
1331
AL117472.1
-
23, 24
2449
AL832353.1
-
4, 5
916
AM260536.1
-
30, 31
3799
BC042612.1
-
23, 24
2790
BC152463.1
-
24, 25
2318
8862599
BX640814.1
-
13, 14
2381
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
muscle contraction
cell-matrix adhesion
insulin receptor signaling pathway
positive regulation of signal transduction
glucose transport
cellular response to insulin stimulus
stress fiber assembly
positive regulation of glycogen biosynthetic process
positive regulation of glucose import
positive regulation of lipid biosynthetic process
focal adhesion assembly
positive regulation of establishment of protein localization to plasma membrane
actin binding
SH3/SH2 adaptor activity
insulin receptor binding
protein binding
cytoskeletal protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-65b4bdbffd-tfbk7:80/10.239.166.103:80.
git-commit: b16971a8df2afcd8df684aafbc9bf1e9f45e42d4
git-url: git@github.com:thermofisher/magellan-core.git
git-branch: origin/release/2.6.0-2024.04.15-1.0