Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
CDMO and Clinical Trial Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other CELF2 SIRNA Assays ›
Gene Symbol:
CELF2
Gene Name:
CUGBP, Elav-like family member 2
Gene Aliases:
BRUNOL3, CELF-2, CUG-BP2, CUGBP2, ETR-3, ETR3, NAPOR
Chromosome Location:
Chr.10: 10462515 - 11336675 on Build GRCh38
UniGene ID:
Hs.309288
Species:
Human
siRNA ID
HSS116449
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: CELF2
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001025076.2
NP_001020247.1
10
1278
NM_001025077.2
NP_001020248.1
10
1135
NM_001083591.1
NP_001077060.1
10
1278
NM_001326317.1
NP_001313246.1
12
1137
NM_001326318.1
NP_001313247.1
12
1137
NM_001326319.1
NP_001313248.1
13
1175
NM_001326320.1
NP_001313249.1
13
1231
NM_001326321.1
NP_001313250.1
12
1137
NM_001326323.1
NP_001313252.1
13
1231
NM_001326324.1
NP_001313253.1
11
1114
NM_001326325.1
NP_001313254.1
12
1208
NM_001326326.1
NP_001313255.1
11
1133
NM_001326327.1
NP_001313256.1
11
1133
NM_001326328.1
NP_001313257.1
12
1227
NM_001326329.1
NP_001313258.1
10
1167
NM_001326330.1
NP_001313259.1
10
1167
NM_001326331.1
NP_001313260.1
10
1319
NM_001326332.1
NP_001313261.1
10
1319
NM_001326333.1
NP_001313262.1
9
1239
NM_001326334.1
NP_001313263.1
11
1504
NM_001326335.1
NP_001313264.1
10
1319
NM_001326336.1
NP_001313265.1
10
1319
NM_001326337.1
NP_001313266.1
10
1319
NM_001326338.1
NP_001313267.1
9
1225
NM_001326339.1
NP_001313268.1
9
1225
NM_001326340.1
NP_001313269.1
10
1231
NM_001326341.1
NP_001313270.1
10
1231
NM_001326342.1
NP_001313271.1
10
1231
NM_001326343.1
NP_001313272.1
10
1231
NM_001326344.1
NP_001313273.1
10
1021
NM_001326345.1
NP_001313274.1
10
1099
NM_001326346.1
NP_001313275.1
8
1264
NM_001326347.1
NP_001313276.1
10
1429
NM_001326348.1
NP_001313277.1
10
1427
NM_001326349.1
NP_001313278.1
10
1427
NM_006561.3
NP_006552.3
10
1224
XM_005252354.4
XP_005252411.1
10
1314
XM_005252357.4
XP_005252414.1
8
933
XM_005252358.3
XP_005252415.1
8
1274
XM_006717373.2
XP_006717436.1
9
1354
XM_011519294.2
XP_011517596.1
10
1314
XM_011519297.1
XP_011517599.1
10
1437
XM_017015542.1
XP_016871031.1
9
990
XM_017015543.1
XP_016871032.1
10
1080
XM_017015544.1
XP_016871033.1
10
1080
XM_017015545.1
XP_016871034.1
10
1080
XM_017015546.1
XP_016871035.1
10
1080
XM_017015547.1
XP_016871036.1
10
1314
XM_017015548.1
XP_016871037.1
10
1080
XM_017015549.1
XP_016871038.1
10
1080
XM_017015550.1
XP_016871039.1
10
2060
XM_017015551.1
XP_016871040.1
10
1314
XM_017015552.1
XP_016871041.1
10
1437
XM_017015553.1
XP_016871042.1
10
1437
XM_017015554.1
XP_016871043.1
10
1437
XM_017015555.1
XP_016871044.1
9
990
XM_017015556.1
XP_016871045.1
9
1354
XM_017015557.1
XP_016871046.1
11
1019
XM_017015558.1
XP_016871047.1
9
1006
XM_017015559.1
XP_016871048.1
9
1354
XM_017015560.1
XP_016871049.1
9
1015
XM_017015561.1
XP_016871050.1
9
1342
XM_017015562.1
XP_016871051.1
9
1354
XM_017015563.1
XP_016871052.1
9
1341
XM_017015564.1
XP_016871053.1
8
930
XM_017015565.1
XP_016871054.1
8
933
XM_017015566.1
XP_016871055.1
8
930
XM_017015567.1
XP_016871056.1
9
1070
XM_017015568.1
XP_016871057.1
11
1057
XM_017015569.1
XP_016871058.1
9
1357
XM_017015570.1
XP_016871059.1
8
933
XM_017015571.1
XP_016871060.1
9
1357
XM_017015572.1
XP_016871061.1
9
1058
GenBank mRNA
AF036956.1
-
10
1194
AF090693.1
-
10
1110
AF090694.1
-
10
1062
AF432906.1
-
10
1130
AK293164.1
-
10
1171
AK295511.1
-
8
865
AK297313.1
-
8
1055
AK297380.1
-
8
933
AK299548.1
-
10
1054
AK299992.1
-
10
1211
AK302681.1
-
7
829
AK314087.1
-
10
1233
AK316354.1
-
10
1030
BC036391.1
-
10
1196
5260623
BM699598.1
-
4
319
U69546.1
-
10
1011
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
RNA processing
mRNA processing
regulation of heart contraction
nucleotide binding
RNA binding
poly(A) RNA binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-65b4bdbffd-d6q6k:80/10.239.161.116:80.
git-commit: b16971a8df2afcd8df684aafbc9bf1e9f45e42d4
git-url: git@github.com:thermofisher/magellan-core.git
git-branch: origin/release/2.6.0-2024.04.15-1.0