Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CXCR6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006564.1 | NP_006555.1 | 2 | 700 | ||
XM_005264809.2 | XP_005264866.1 | 2 | 774 | |||
XM_011533290.2 | XP_011531592.1 | 2 | 1299 | |||
XM_011533291.2 | XP_011531593.1 | 2 | 1250 | |||
GenBank mRNA | AF007545.1 | - | 1 | 619 | ||
AK313754.1 | - | 2 | 774 | |||
BC033584.1 | - | 1 | 651 | 5225727 | ||
JC209412.1 | - | 2 | 700 | |||
U73529.1 | - | 1 | 649 | |||
U73531.1 | - | 1 | 2081 | |||
Y13248.1 | - | 2 | 700 |