Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: SLC12A7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006598.2 | NP_006589.2 | 5 | 578 | ||
XM_005248231.3 | XP_005248288.1 | 5 | 583 | |||
XM_011513939.2 | XP_011512241.1 | 5 | 672 | |||
XM_011513940.2 | XP_011512242.1 | 5 | 507 | |||
XM_011513941.2 | XP_011512243.2 | 5 | 602 | |||
XM_017008958.1 | XP_016864447.1 | 5 | 882 | |||
XM_017008959.1 | XP_016864448.1 | 5 | 534 | |||
GenBank mRNA | AF105365.1 | - | 5 | 516 | ||
AK302856.1 | - | 5 | 556 | |||
BC007760.2 | - | 5 | 558 | |||
BC018982.2 | - | 5 | 558 | |||
BC098390.1 | - | 5 | 556 | 5265743 |