Search
Search

KIF1C| Gene Symbol: KIF1C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_006612.5 | NP_006603.2 | 8 | 1017 | ||
| XM_005256424.2 | XP_005256481.1 | 9 | 1008 | |||
| GenBank mRNA | AB014606.2 | - | 8 | 1013 | ||
| AK307401.1 | - | 8 | 860 | |||
| BC034993.1 | - | 8 | 991 | 5266798 | ||
| U91329.1 | - | 7 | 773 | |||