Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other HSPH1 SIRNA Assays ›
          Gene Symbol:
          HSPH1
          Gene Name:
          heat shock protein family H (Hsp110) member 1
          Gene Aliases:
          HSP105, HSP105A, HSP105B, NY-CO-25
          Chromosome Location:
          Chr.13: 31134974 - 31163109 on Build GRCh38
          UniGene ID:
          Hs.743267
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID HSS116686
          Size 20 nmol
          Availability Made To Order
          Catalog # 1299001
          Price Contact Us
          Your Price
          Online offer:
          3062.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: HSPH1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001286503.1 NP_001273432.1 8 1371
          NM_001286504.1 NP_001273433.1 8 1232
          NM_001286505.1 NP_001273434.1 6 998
          NM_006644.3 NP_006635.2 8 1371
          XM_005266236.1 XP_005266293.1 8 1953
          XM_011534887.2 XP_011533189.1 8 1953
          XM_011534888.1 XP_011533190.1 6 1061
          XM_017020361.1 XP_016875850.1 8 1295
          XM_017020362.1 XP_016875851.1 8 1953
          XM_017020363.1 XP_016875852.1 8 1295
          XM_017020364.1 XP_016875853.1 6 1061
          GenBank mRNA AB003333.1 - 8 1245
          AB003334.1 - 8 1277
          AF039695.1 - 8 974
          AK293955.1 - 8 1247
          AK302294.1 - 6 649
          AK302430.1 - 8 1232
          AK302832.1 - 10 1322
          AK303029.1 - 7 749
          AK314576.1 - 8 1295
          BC037553.1 - 8 1273 4540927
          BX648125.1 - 8 1209
          BX648623.1 - 2 1579
          D86956.1 - 8 1279

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          receptor-mediated endocytosis
          response to unfolded protein
          positive regulation of MHC class I biosynthetic process
          positive regulation of transcription from RNA polymerase II promoter
          chaperone mediated protein folding requiring cofactor
          positive regulation of NK T cell activation
          positive regulation of protein tyrosine kinase activity
          regulation of cellular response to heat
          negative regulation of establishment of protein localization to mitochondrion
          negative regulation of intrinsic apoptotic signaling pathway in response to hydrogen peroxide
          negative regulation of p38MAPK cascade
          adenyl-nucleotide exchange factor activity
          protein binding
          ATP binding
          alpha-tubulin binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-lffdw:80/100.66.79.31:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0