Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM114A2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001317993.1 | NP_001304922.1 | 11 | 1173 | ||
NM_001317994.1 | NP_001304923.1 | 10 | 1690 | |||
NM_001317995.1 | NP_001304924.1 | 9 | 916 | |||
NM_018691.3 | NP_061161.2 | 10 | 1126 | |||
XM_005268360.4 | XP_005268417.1 | 11 | 1251 | |||
XM_005268361.4 | XP_005268418.1 | 10 | 1154 | |||
XM_011537540.2 | XP_011535842.1 | 10 | 1480 | |||
XM_011537541.2 | XP_011535843.1 | 10 | 2062 | |||
XM_011537542.2 | XP_011535844.1 | 10 | 1351 | |||
XM_017008967.1 | XP_016864456.1 | 10 | 1145 | |||
XM_017008968.1 | XP_016864457.1 | 10 | 1167 | |||
GenBank mRNA | AF159700.1 | - | 10 | 1619 | ||
AK024664.1 | - | 10 | 1091 | |||
AK095446.1 | - | 10 | 1046 | |||
AK225821.1 | - | 10 | 1109 | |||
AK298943.1 | - | 9 | 885 | |||
AK303865.1 | - | 9 | 966 | |||
AK313331.1 | - | 10 | 1098 | |||
BC011414.1 | - | 11 | 1108 | 3855873 | ||
BC050701.1 | - | 10 | 1086 |