Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PPARGC1A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013261.3 | NP_037393.1 | 8 | 1391 | ||
XM_005248131.4 | XP_005248188.1 | 8 | 2134 | |||
XM_005248132.1 | XP_005248189.1 | 8 | 1313 | |||
XM_005248134.4 | XP_005248191.1 | 8 | 1380 | |||
XM_011513765.2 | XP_011512067.1 | 9 | 1883 | |||
XM_011513766.1 | XP_011512068.1 | 9 | 1465 | |||
XM_011513767.2 | XP_011512069.1 | 9 | 3603 | |||
XM_011513768.1 | XP_011512070.1 | 8 | 1364 | |||
XM_011513769.2 | XP_011512071.1 | 8 | 1380 | |||
XM_011513770.2 | XP_011512072.1 | 7 | 1601 | |||
XM_011513771.1 | XP_011512073.1 | 7 | 1210 | |||
XM_017007664.1 | XP_016863153.1 | 10 | 1840 | |||
GenBank mRNA | AB061325.1 | - | 8 | 1377 | ||
AF106698.1 | - | 8 | 1391 | |||
AF159714.1 | - | 8 | 1300 | |||
AF186379.1 | - | 8 | 1310 | |||
HQ695733.1 | - | 7 | 1601 | |||
JQ772116.1 | - | 9 | 1679 | |||
JQ772117.1 | - | 9 | 1782 | |||
JQ772118.1 | - | 10 | 1878 | |||
JQ772119.1 | - | 10 | 1883 | |||
JQ772120.1 | - | 10 | 1965 |