Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
還沒有帳戶嗎 ?
立即註冊
產品
Antibodies
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Cell Culture Media
Chemicals
Western Blot Products
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Greener Products
查看所有產品類別
應用
生物製程
細胞培養和轉染
食品和飲料
實驗室解決方案
製藥和生物製藥
即時定量PCR
半導體分析
臨床和診斷
數位解決方案
分子光譜、同位素與元素分析
查看所有應用與技術
服務
360° CDMO 和 CRO 服務
CDMO 服务
临床试验 CRO 服务
客製化服務
企業服務
金融與租賃服務
儀器服務
實驗室資訊學
授權與商業供應
培訓服務
查看所有服務
支援
線上訂購協助
數位解決方案
產品支持
技術相關資訊
培訓與教育
查看所有協助與支援
熱門商品
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
特價商品
聯絡我們
快速訂購
訂單狀態與貨物追蹤
文件和證書
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
訂單查詢
快速訂購
登入
登入
還沒有帳戶嗎 ?
立即註冊
帳戶
訂單
Connect: Lab, Data, Apps
客製化產品與項目
儀器管理
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other BRD8 SIRNA Assays ›
Gene Symbol:
BRD8
Gene Name:
bromodomain containing 8
Gene Aliases:
SMAP, SMAP2, p120
Chromosome Location:
Chr.5: 138139766 - 138178669 on Build GRCh38
UniGene ID:
Hs.519337
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
HSS116822
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Contact Us
Your Price
Online offer:
3062.0
Check your price ›
新增到購物車
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: BRD8
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001164326.1
NP_001157798.1
13
1811
NM_001300961.1
NP_001287890.1
13
1807
NM_001300962.1
NP_001287891.1
12
1732
NM_001300966.1
NP_001287895.1
14
1899
NM_006696.3
NP_006687.3
14
2021
NM_139199.1
NP_631938.1
13
1802
XM_005271855.2
XP_005271912.1
14
2012
XM_005271856.2
XP_005271913.1
14
2012
XM_005271857.2
XP_005271914.1
13
1802
XM_005271859.3
XP_005271916.1
12
1583
XM_005271860.3
XP_005271917.1
14
2012
XM_005271861.3
XP_005271918.1
13
1793
XM_011543109.1
XP_011541411.1
14
2012
XM_011543110.1
XP_011541412.1
14
1998
XM_017008969.1
XP_016864458.1
13
1793
XM_017008970.1
XP_016864459.1
14
2012
XM_017008971.1
XP_016864460.1
14
2012
XM_017008972.1
XP_016864461.1
13
1802
XM_017008973.1
XP_016864462.1
13
1793
XM_017008974.1
XP_016864463.1
13
1802
XM_017008975.1
XP_016864464.1
13
1793
XM_017008976.1
XP_016864465.1
12
1583
GenBank mRNA
AB209079.1
-
14
1967
AF016270.1
-
13
1953
AK225928.1
-
13
1774
AK289951.1
-
14
1981
AK293621.1
-
12
1692
AK297608.1
-
4
1090
AK297756.1
-
13
1767
AK312333.1
-
14
1981
AL136823.1
-
13
1775
BC008039.2
-
14
1963
3009915
BC008076.2
-
14
1963
3009915
X87613.1
-
13
1829
X94234.1
-
12
1741
Back To Top
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Rat: NM_001008509, XM_006254564, XM_008772029, XM_006254562, XM_006254563, and others.
Gene Ontology Categories:
Function(s)
Process(es)
transcription, DNA-templated
regulation of transcription, DNA-templated
regulation of transcription from RNA polymerase II promoter
signal transduction
cell surface receptor signaling pathway
intracellular receptor signaling pathway
regulation of growth
histone H4 acetylation
histone H2A acetylation
transcription factor activity, sequence-specific DNA binding
thyroid hormone receptor activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-8jdkq:80/100.66.75.107:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0