Search
Search

SUGT1| Gene Symbol: SUGT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001130912.2 | NP_001124384.1 | 14 | 1260 | ||
| NM_001320831.1 | NP_001307760.1 | 14 | 1292 | |||
| NM_006704.4 | NP_006695.1 | 13 | 1164 | |||
| XM_011534891.2 | XP_011533193.1 | 14 | 1355 | |||
| XM_011534893.2 | XP_011533195.1 | 13 | 1073 | |||
| XM_017020365.1 | XP_016875854.1 | 14 | 1245 | |||
| XM_017020366.1 | XP_016875855.1 | 14 | 1120 | |||
| XM_017020367.1 | XP_016875856.1 | 14 | 1154 | |||
| XM_017020368.1 | XP_016875857.1 | 14 | 1166 | |||
| GenBank mRNA | AF132856.1 | - | 13 | 934 | ||
| AJ344097.1 | - | 13 | 1027 | |||
| AK291402.1 | - | 14 | 1077 | |||
| AK292128.1 | - | 13 | 1024 | |||
| AK302363.1 | - | 14 | 1139 | |||
| AY271314.1 | - | 14 | 1097 | |||
| AY321358.1 | - | 14 | 1112 | |||
| BC000911.1 | - | 13 | 989 | 2985858 | ||
| BT009798.1 | - | 13 | 934 | |||