Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EDAR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022336.3 | NP_071731.1 | 8, 9 | 1171 | ||
XM_006712204.1 | XP_006712267.1 | 7, 8 | 1151 | |||
XM_011510502.2 | XP_011508804.2 | 7, 8 | 1030 | |||
XM_011510503.2 | XP_011508805.2 | 8, 9 | 927 | |||
GenBank mRNA | AF130988.1 | - | 8, 9 | 1172 |