Search
Search

EDAR| Gene Symbol: EDAR | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022336.3 | NP_071731.1 | 12 | 1610 | ||
| XM_006712204.1 | XP_006712267.1 | 11 | 1590 | |||
| XM_011510502.2 | XP_011508804.2 | 11 | 1469 | |||
| XM_011510503.2 | XP_011508805.2 | 12 | 1366 | |||
| GenBank mRNA | AF130988.1 | - | 12 | 1611 | ||
| AK296936.1 | - | 11 | 1500 | |||
| AK313781.1 | - | 12 | 1494 | |||
| BC093870.1 | - | 11 | 1202 | 7939715 | ||
| BC093872.1 | - | 11 | 1202 | 7939717 | ||
| CR749352.1 | - | 12 | 1719 | |||