Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZPBP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001159878.1 | NP_001153350.1 | 2 | 239 | ||
NM_007009.2 | NP_008940.2 | 2 | 239 | |||
XM_011515095.2 | XP_011513397.1 | 2 | 233 | |||
XM_011515096.2 | XP_011513398.1 | 2 | 211 | |||
XM_011515097.1 | XP_011513399.1 | 2 | 233 | |||
XM_011515100.2 | XP_011513402.1 | 2 | 187 | |||
XM_011515101.2 | XP_011513403.1 | 2 | 187 | |||
XM_011515102.2 | XP_011513404.1 | 2 | 412 | |||
XM_011515103.1 | XP_011513405.1 | 2 | 233 | |||
XM_017011707.1 | XP_016867196.1 | 2 | 233 | |||
GenBank mRNA | AK312799.1 | - | 2 | 211 | ||
BC005223.1 | - | 2 | 198 | 3950015 | ||
BT006822.1 | - | 2 | 169 | |||
D17570.1 | - | 2 | 205 |