Search
Search

TPPP| Gene Symbol: TPPP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_007030.2 | NP_008961.1 | 2 | 215 | ||
| XM_005248237.3 | XP_005248294.2 | 2 | 280 | |||
| XM_017008993.1 | XP_016864482.1 | 2 | 2411 | |||
| XM_017008994.1 | XP_016864483.1 | 4 | 266 | |||
| GenBank mRNA | AB017016.1 | - | 2 | 126 | ||
| AK054920.1 | - | 1 | 812 | |||
| AK098392.1 | - | 2 | 171 | |||
| AY319261.1 | - | 1 | 93 | |||
| BC040496.1 | - | 2 | 139 | |||
| BC131506.1 | - | 1 | 111 | 40082752 | ||
| BC152382.1 | - | 1 | 96 | 100013441 | ||
| BT020035.1 | - | 1 | 93 | |||
| DB478250.2 | - | 2 | 215 | |||