Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FICD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007076.2 | NP_009007.2 | 3 | 614 | ||
GenBank mRNA | AK000014.1 | - | 3 | 588 | ||
AK026074.1 | - | 3 | 614 | |||
AY358992.1 | - | 3 | 583 | |||
BC001342.1 | - | 3 | 572 | 3462741 |