Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BAZ1A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013448.2 | NP_038476.2 | 18 | 3086 | ||
NM_182648.1 | NP_872589.1 | 17 | 2990 | |||
XM_011536374.1 | XP_011534676.1 | 17 | 2385 | |||
XM_011536376.2 | XP_011534678.2 | 8 | 1213 | |||
XM_017020942.1 | XP_016876431.1 | 7 | 1116 | |||
GenBank mRNA | AB032252.1 | - | 17 | 2987 | ||
AB209255.1 | - | 9 | 1337 | |||
AF213467.1 | - | 18 | 2684 | |||
AF221130.1 | - | 17 | 2518 | |||
AL050089.1 | - | 1 | 283 |