Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SYNPO | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109974.2 | NP_001103444.1 | 2 | 927 | ||
NM_001166208.1 | NP_001159680.1 | 3 | 1491 | |||
NM_001166209.1 | NP_001159681.1 | 3 | 1424 | |||
NM_007286.5 | NP_009217.3 | 2 | 927 | |||
XM_005268370.1 | XP_005268427.1 | 2 | 870 | |||
XM_005268371.1 | XP_005268428.1 | 2 | 790 | |||
XM_006714755.3 | XP_006714818.1 | 3 | 1479 | |||
XM_011537552.2 | XP_011535854.1 | 3 | 1476 | |||
XM_017009007.1 | XP_016864496.1 | 3 | 1512 | |||
XM_017009008.1 | XP_016864497.1 | 3 | 1451 | |||
GenBank mRNA | AB028952.1 | - | 2 | 881 | ||
AF499136.1 | - | 2 | 841 | |||
AF499137.1 | - | 2 | 841 | |||
AK127049.1 | - | 3 | 1424 | |||
AK290178.1 | - | 2 | 910 | |||
AL831818.1 | - | 3 | 1491 | |||
BC142683.1 | - | 2 | 848 | 40148430 | ||
BC146665.1 | - | 2 | 894 | 40148077 | ||
BC152538.1 | - | 2 | 881 | |||
Y11072.1 | - | 1 | 385 |