Search
Search

NID2| Gene Symbol: NID2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_007361.3 | NP_031387.3 | 11, 12 | 2753 | ||
| XM_005267405.4 | XP_005267462.1 | 10, 11 | 2837 | |||
| GenBank mRNA | AJ223500.1 | - | 11, 12 | 2519 | ||
| AK291652.1 | - | 11, 12 | 2581 | |||
| BC035608.1 | - | 7, 8 | 1355 | 5205293 | ||
| BX648241.1 | - | 12, 13 | 2526 | |||
| D86425.1 | - | 11, 12 | 2522 | |||