Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZFP30 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001320666.1 | NP_001307595.1 | 6 | 1046 | ||
NM_001320667.1 | NP_001307596.1 | 6 | 952 | |||
NM_001320668.1 | NP_001307597.1 | 6 | 822 | |||
NM_001320669.1 | NP_001307598.1 | 6 | 760 | |||
NM_001320670.1 | NP_001307599.1 | 6 | 757 | |||
NM_001320671.1 | NP_001307600.1 | 6 | 663 | |||
NM_014898.2 | NP_055713.1 | 6 | 993 | |||
GenBank mRNA | AB023178.1 | - | 6 | 993 | ||
AK297301.1 | - | 6 | 631 | |||
BC041087.1 | - | 6 | 1022 | 5579247 | ||
BC050542.1 | - | 6 | 951 | |||
GU388430.1 | - | 6 | 822 |