Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PPM1E | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014906.4 | NP_055721.3 | 3, 4 | 922 | ||
NR_048561.1 | 2, 3 | 722 | ||||
XM_005257169.4 | XP_005257226.1 | 2, 3 | 147 | |||
XM_011524534.2 | XP_011522836.1 | 2, 3 | 1043 | |||
XM_017024359.1 | XP_016879848.1 | 3, 4 | 1439 | |||
GenBank mRNA | AB028995.1 | - | 3, 4 | 926 | ||
AF260269.1 | - | 3, 4 | 953 | |||
AF520614.1 | - | 3, 4 | 799 | |||
AK289966.1 | - | 3, 4 | 928 | |||
BC136290.1 | - | 3, 4 | 868 | 9020277 | ||
BC143845.1 | - | 2, 3 | 668 | |||
BC151228.1 | - | 3, 4 | 926 | 9007247 | ||
CR749253.1 | - | 2, 3 | 162 |
Gene Symbol: TRIM37 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017024663.1 | XP_016880152.1 | 25 | 3731 |