Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RAB18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256410.1 | NP_001243339.1 | 8 | 844 | ||
NM_001256411.1 | NP_001243340.1 | 6 | 690 | |||
NM_001256412.1 | NP_001243341.1 | 5 | 565 | |||
NM_001256415.1 | NP_001243344.1 | 6 | 685 | |||
NM_021252.4 | NP_067075.1 | 7 | 757 | |||
NR_046172.1 | 6 | 695 | ||||
GenBank mRNA | AB209698.1 | - | 3 | 5519 | ||
AF087860.1 | - | 7 | 615 | |||
AF136974.1 | - | 7 | 671 | |||
AF137372.1 | - | 7 | 561 | |||
AF498950.1 | - | 7 | 561 | |||
AK001555.1 | - | 7 | 561 | |||
AK223153.1 | - | 7 | 584 | |||
AK295443.1 | - | 5 | 435 | |||
AK297629.1 | - | 6 | 560 | |||
AK299457.1 | - | 6 | 561 | |||
AL136734.1 | - | 7 | 610 | |||
AY574034.1 | - | 8 | 733 | |||
BC015014.2 | - | 7 | 611 | 3922029 | ||
BC029350.1 | - | 7 | 627 | 4819866 | ||
BP210053.1 | - | 6 | 559 | |||
BT009840.1 | - | 7 | 561 | |||
CR533455.1 | - | 7 | 561 |