Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
CDMO and Clinical Trial Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other LIMCH1 SIRNA Assays ›
Gene Symbol:
LIMCH1
Gene Name:
LIM and calponin homology domains 1
Gene Aliases:
LIMCH1A, LMO7B
Chromosome Location:
Chr.4: 41359607 - 41700044 on Build GRCh38
UniGene ID:
Hs.335163
Species:
Human
siRNA ID
HSS118005
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: LIMCH1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001112717.2
NP_001106188.1
25
3147
NM_001112718.2
NP_001106189.1
25
3144
NM_001112719.2
NP_001106190.1
20
2929
NM_001112720.2
NP_001106191.1
19
2893
NM_001289122.1
NP_001276051.1
25
3183
NM_001289124.1
NP_001276053.1
25
3228
NM_014988.3
NP_055803.2
26
3225
XM_005248057.1
XP_005248114.1
33
4866
XM_005248058.1
XP_005248115.1
33
4863
XM_005248060.1
XP_005248117.1
33
4854
XM_005248061.1
XP_005248118.1
32
4782
XM_005248067.2
XP_005248124.1
31
5728
XM_005248072.1
XP_005248129.1
26
3222
XM_005248074.1
XP_005248131.1
25
3186
XM_005248075.1
XP_005248132.1
24
3108
XM_006713996.1
XP_006714059.1
32
4776
XM_011513642.1
XP_011511944.1
34
4941
XM_011513643.1
XP_011511945.1
34
4938
XM_011513644.1
XP_011511946.1
34
4932
XM_011513645.1
XP_011511947.1
33
4857
XM_011513646.1
XP_011511948.1
33
4854
XM_011513648.1
XP_011511950.1
33
4698
XM_011513649.1
XP_011511951.1
29
4600
XM_011513651.2
XP_011511953.1
32
5820
XM_011513653.1
XP_011511955.1
27
3309
XM_011513654.1
XP_011511956.1
26
3273
XM_011513655.1
XP_011511957.1
21
2932
XM_011513656.1
XP_011511958.1
21
2890
XM_011513657.1
XP_011511959.1
20
2845
XM_011513658.2
XP_011511960.1
24
3369
XM_017007894.1
XP_016863383.1
34
4929
XM_017007895.1
XP_016863384.1
33
4857
XM_017007896.1
XP_016863385.1
32
4770
XM_017007897.1
XP_016863386.1
31
4695
XM_017007898.1
XP_016863387.1
26
3234
XM_017007899.1
XP_016863388.1
26
3231
XM_017007900.1
XP_016863389.1
25
3150
XM_017007901.1
XP_016863390.1
25
3141
XM_017007902.1
XP_016863391.1
24
3063
XM_017007903.1
XP_016863392.1
22
2968
XM_017007904.1
XP_016863393.1
21
2881
XM_017007905.1
XP_016863394.1
20
2806
GenBank mRNA
AB029025.2
-
26
3223
AK027231.1
-
12
1149
AK294774.1
-
20
3034
AK295784.1
-
19
2560
AK295836.1
-
20
2670
AK299297.1
-
19
2763
AK299835.1
-
20
2705
AK302674.1
-
12
1395
AK309846.1
-
19
2835
AL831962.2
-
31
4609
BC023546.2
-
22
2997
4110496
BC029735.1
-
10
899
BC095394.1
-
15
1444
BX537916.1
-
19
2763
BX640692.1
-
18
2521
CR749205.1
-
20
2799
CR936601.1
-
25
3154
CR936610.1
-
25
3229
CR936658.1
-
22
2895
CR936661.1
-
24
3048
CR936664.1
-
25
3145
Back To Top
More Information
Panther Classification:
Molecular Function -
actin or actin-binding cytoskeletal protein
Gene Ontology Categories:
Function(s)
Process(es)
actomyosin structure organization
actin binding
zinc ion binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-5c7f9f6c68-kz7s5:80/10.239.165.170:80.
git-commit: b16971a8df2afcd8df684aafbc9bf1e9f45e42d4
git-url: git@github.com:thermofisher/magellan-core.git
git-branch: origin/release/2.6.0-2024.04.15-1.0