Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Cell Analysis
Antibodies
Mass Spectrometry
Cell Culture
Laboratory Instruments
Clinical and Diagnostics
Chromatography
Laboratory Equipment
Laboratory Supplies
Molecular Biology and Nucleic Acid Analysis
Sequence-Specific Nucleic Acid Products
See all product categories
Applications
Cell Culture and Transfection
Flow Cytometry
Cancer Research
Chromatography
Sequencing
PCR
Lab Solutions
Allergy Diagnostics
See all applications and techniques
Services
Cell Biology Services
Custom Services
Training Services
Enterprise Services
Lab Informatics Services
Financial and Leasing Services
Partnering and Licensing Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Food Safety Inspection Services
See all services
Help and Support
How to Order
Promotions and Online Offers
Contact Us
Change Location
Create a New Account
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Special Offers
Contact Us
Quick Order
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Contact Us
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other TMCC1 SIRNA Assays ›
Gene Symbol:
TMCC1
Gene Name:
transmembrane and coiled-coil domain family 1
Gene Aliases:
-
Chromosome Location:
Chr.3: 129647792 - 129893605 on Build GRCh38
UniGene ID:
Hs.477547
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
HSS118048
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: TMCC1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001017395.3
NP_001017395.2
4
1649
NM_001128224.2
NP_001121696.1
4
1050
NR_033361.1
2
1195
XM_006713542.3
XP_006713605.1
5
1762
XM_006713543.3
XP_006713606.1
5
1759
XM_006713544.1
XP_006713607.1
5
1887
XM_006713550.3
XP_006713613.1
5
1320
XM_006713552.3
XP_006713615.1
2
942
XM_006713553.3
XP_006713616.1
2
2234
XM_011512572.2
XP_011510874.1
6
1922
XM_011512573.2
XP_011510875.1
5
2501
XM_011512576.2
XP_011510878.1
5
1755
XM_011512579.2
XP_011510881.1
4
1510
XM_011512580.2
XP_011510882.1
7
1647
XM_011512581.2
XP_011510883.1
6
1484
XM_011512582.1
XP_011510884.1
2
1276
XM_011512583.2
XP_011510885.1
4
1053
XM_011512584.2
XP_011510886.1
6
1372
XM_017005931.1
XP_016861420.1
5
1670
XM_017005932.1
XP_016861421.1
6
1909
XM_017005933.1
XP_016861422.1
4
1706
XM_017005934.1
XP_016861423.1
8
1720
XM_017005935.1
XP_016861424.1
9
1809
XM_017005936.1
XP_016861425.1
6
1483
XM_017005937.1
XP_016861426.1
6
1417
XM_017005938.1
XP_016861427.1
8
1783
XM_017005939.1
XP_016861428.1
7
1677
XM_017005940.1
XP_016861429.1
4
1055
XM_017005941.1
XP_016861430.1
4
1181
GenBank mRNA
AB018322.1
-
1
216
AK124671.1
-
2
1196
AK291448.1
-
4
1468
AK293408.1
-
2
732
BC030619.2
-
2
1113
BC039859.1
-
4
1556
5527623
BC047517.1
-
2
1113
BX640712.1
-
2
703
CR749206.1
-
2
1175
Back To Top
More Information
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-ctdjb:80/100.66.76.55:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0